Learn More
NHE-1 (Na+/H+ Exchangers) Mouse, Unlabeled, Clone: 54, BD
Mouse Monoclonal Antibody
401.00€
Specifications
Antigen | NHE-1 (Na+/H+ Exchangers) |
---|---|
Clone | 54 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15806029
|
BD Biosciences
611775 |
150 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
The extrusion of H+ in exchange for extracellular Na+ is important for many cellular processes, such as pH homeostasis, volume regulation, and transepithelial ion and water transport. Na+/H+ Exchangers (NHE) are integral membrane proteins that mediate electroneutral exchange of one Na+ ion for one H+ ion. Six NHE forms, NHE-1 thru -6, have been identified. NHE-1 and NHE-6 are widely expressed, while the other NHE forms have restricted expression. The common structure of all NHE forms includes 10-12 N-terminal membrane (M) spanning regions, a conserved M6 and M7 region that may participate in ion transport, and a large C-terminal cytoplasmic region that may be involved in the regulation of ion exchange activity. NHE-1, for example, contains 12 M regions plus domains for volume sensitivity, calmodulin-binding, CHP-binding, and PKC phosphorylation in the cytoplasmic region. Regulation of NHE-1 ion exchange activity may occur through phosphoinositide binding, as well as PKC- and PKA-dependent signaling pathways. Mutation of NHE-1 in mice causes neuronal death in the cerebellum and brainstem, leading to ataxia and seizures. Thus, NHE-1 is a ubiquitous NHE that is essential for normal brain function.
Although this antibody was developed against the NHE-1 antigen, investigators should note that crossreactivity to other NHE isoforms or variants may be possible.
Although this antibody was developed against the NHE-1 antigen, investigators should note that crossreactivity to other NHE isoforms or variants may be possible.
Specifications
NHE-1 (Na+/H+ Exchangers) | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Na+/H+ Exchangers |
54 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Rat NHE-1 aa. 682-801 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.