Learn More
anti-NOP16 Matched Antibody Pair, Ab-2, Abnova
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-NOP16 (100μg); Mouse purified polyclonal anti-NOP16 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
NOP16 is transcriptionally regulated by c-Myc (MYC; MIM 190080), upregulated in breast cancer, and overexpression is associated with poor patient survival.
ELISA
Specifications
Specifications
| Antigen | NOP16 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NOP16. |
| Gene | NOP16 |
| Gene Alias | HSPC111/HSPC185 |
| Gene Symbols | NOP16 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.