missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR1M1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSR00259W
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 100 μL of distilled water.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Specifications
OR1M1 | |
Polyclonal | |
OR1M1 | |
Olfactory receptor, family 1, subfamily M, member 1, Olfactory receptor 19-6, OR19-6, Olfactory receptor 1M1, OR19-5 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8NGA1 | |
OR1M1 | |
A synthetic peptide from human olfactory receptor OR1M1 | |
RUO | |
125963 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |