missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MAP2K4 (Thr261) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517878
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the corresponding phosphorylated residues of MEK1, MEK2 or MKK3.
Phospho-MAP2K4 (Thr261) Polyclonal antibody specifically detects Phospho-MAP2K4 (Thr261) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-MAP2K4 (Thr261) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P45985, P47809 | |
MAP2K4 | |
Synthetic phosphopeptide corresponding to residues around pThr261 of human SEK1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MAP2K4 | |
C-JUN N-terminal kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 2; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4a; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; JNKK1; MAP kinase activator 2; MAP kinase kinase 4; Map2k4; map2k4.L; map2k4a; map2k4-a; map2k4-b; MAP2K4delta; MAPK/ERK kinase 4; MAPKK 4; MAPKK4; MEK 4; MEK4; mitogen activated protein kinase kinase 4; mitogen activated protein kinase kinase 4, variant 1; mitogen-activated protein kinase kinase 4; mitogen-activated protein kinase kinase 4 L homeolog; mitogen-activated protein kinase kinase 4a; MKK4; mkk4a; MKK4A-l; PRKMK4; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SAPKK1; SAPKK-1; Sek1; Serk1; Skk1; Stress-activated protein kinase kinase 1; XELAEV_18044157mg; XMEK2; zgc:136500; zMKK4 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26398, 287398, 6416 | |
-20°C | |
Liquid |