missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p23 (Ser113) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538337
Additional Details : Weight : 0.00900kg
Description
Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes.Specifications
Phospho-p23 (Ser113) | |
Polyclonal | |
Unconjugated | |
PTGES3 | |
P23, TEBP, cPGES, Hsp90 co-chaperone, cytosolic prostaglandin E synthase, cytosolic prostaglandin E2 synthase, progesterone receptor complex p23, prostaglandin E synthase 3, telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10728, 362809, 56351 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15185, Q9R0Q7, P83868 | |
PTGES3 | |
A synthetic phosphopeptide derived from human TEBP around the phosphorylation site of Ser113 (D-D-SP-D-E) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |