missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIKFYVE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513977
Additional Details : Weight : 0.01000kg
Description
PIKFYVE Polyclonal antibody specifically detects PIKFYVE in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
PIKFYVE | |
Polyclonal | |
Unconjugated | |
PIKFYVE | |
1-phosphatidylinositol 3-phosphate 5-kinase; 1-phosphatidylinositol-3-phosphate 5-kinase; 5230400C17Rik; CFD; epididymis luminal protein 37; FAB1; FYVE finger-containing phosphoinositide kinase; HEL37; Kiaa0981; p235; Phosphatidylinositol 3-phosphate 5-kinase; Phosphatidylinositol 3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate/phosphatidylinositol 5-kinase, type III; phosphatidylinositol-4-phosphate 5-kinase, type III; phosphoinositide kinase, FYVE finger containing; phosphoinositide kinase, FYVE-type zinc finger containing; PIKfyve; Pip5k; PIP5K3; Pipk5k3; PipkIII; PIPkin-III; PtdIns(4)P-5-kinase; type III PIP kinase; ZFYVE29; zinc finger, FYVE domain containing 29 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
200576 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q9Y2I7 | |
PIKFYVE | |
KLH conjugated synthetic peptide between 1531-1561 amino acids from the central region of human PIP5K3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |