Learn More
Brady™ Anti-skid Laminated Polyester Tape with Stripes
Product Code. 11751645
Click to view available options
Dimensions (L x W):
18.29 m x 101.6 mm
18.29 m x 152.40 mm
18.29 m x 25.40 mm
18.29 m x 76.20 mm
Unit Size:
1 roll
Product Code. 11751645
Brand: Brady™ 078146
Only null left
Add to basket
Add to basket
Anti-Skid (anti-slip) Laminated Polyester Tape with Stripes helps to prevent slipping and falling on wet or slippery floors.
  • The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit.
  • Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors.
  • Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs.
Specifications
Alternate Part Number STRIPED ANTI-SKID TAPE
Certifications/Compliance ANSI A1264.2-2006, NSFI Certified for High Traction, OSHA 1910.144, OSHA 1910.22
Dimensions (L x W) 18.29 m x 25.40 mm
For Use With (Application) Traction on steps, slick floors, and egress pathway marking
Length (English) 60 ft.
Length (Metric) 18.29 m
Packaging Quantity 1 Roll
Temperature Range (English) -40°F to +158°F
Temperature Range (Metric) -40°C to +70°C
Thickness 0.162 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Adhesive Type Permanent Acrylic
Color Black and Yellow Striped
Material Polyester
Product Type Striped Anti-skid Tape
Type Anti-skid Tape