Learn More
Merck Millipore anti-Synuclein, α Ab-4; Polyclonal
Brand:  Merck Millipore AB5038
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12778786
  • 643.00€ / 50µL
Estimated Shipment: 27-07-2021
Add to basket
Description
Description
α -Synuclein is a small (140-amino acid) cytoplasmic protein of unclear function that is enriched in presynaptic terminals and is the precursor protein of a nonamyloid β component of senile plaques in Alzeheimer's Disease (AD). α -Synuclein is located predominantly in the presynaptic nerve terminals in the brain but is also found in low concentrations in all tissues except liver. Recent studies have shown that abnormal aggregation and accumulation of synaptic proteins, including alpha-Synuclein might be associated with plaque formation in AD and seems to be a major component of Lewy bodies in dementia with Lewy bodies (DLB). It has also been identified in the Glial Cytoplasmic Inclusions (GCIs) found in Multiple System Atrophy (MSA).
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Synuclein, α Ab-4
Serum
Human α-synuclein residues 111-131 coupled to KLH. The peptide sHuence is N-GILEDMPVDPDNEAYEMPSEEC-C.
RUO
Primary
Human, Rat
Unconjugated
Rabbit
50μL
Neuroscience