missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537075
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 48 kDa. Purity is >95% by SDS-PAGE.
TEF1 Polyclonal antibody specifically detects TEF1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TEF1 | |
Polyclonal | |
Unconjugated | |
Tead1 | |
2610024B07Rik; AA; B230114H05Rik; Gtrgeo5; mTEF-1; N TEAD1; NTEF1; NTEF-1; protein GT-IIC; REF1; Tcf13; TCF-13; TEA domain family member 1; TEA domain family member 1 (SV40 transcriptional enhancer factor); TEA domain transcription factor 1; TEAD1; TEAD-1; TEF1; TEF-1; Transcription factor 13; transcriptional enhancer factor 1; transcriptional enhancer factor TEF-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21676, 361630, 7003 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P28347, P30051 | |
Tead1 | |
Synthetic peptide corresponding to amino acids 105-150 of human TEF-1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |