missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEFM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546120
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: NLESLMNVPL FKYKSTVQVC NSILCPKTGR EKRKSPENRF LRKLLKPDIE Sequence homology: Human: 100%
TEFM Polyclonal antibody specifically detects TEFM in Human samples. It is validated for Western Blot.Specifications
TEFM | |
Polyclonal | |
Unconjugated | |
TEFM | |
1110002N22Rik; C17orf42; C9H17orf42; RGD1306595; Tefm; transcription elongation factor of mitochondria; transcription elongation factor, mitochondrial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79736 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96QE5 | |
TEFM | |
Synthetic peptide directed towards the N-terminal of human TEFM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |