missing translation for 'onlineSavingsMsg'
Learn More

Tenascin C Monoclonal Antibody (BC-24), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA126779

Additional Details : Weight : 0.01000kg

Product Code. 11559552

  • 459.00€ / 100µL
Estimated Shipment: 15-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tenascin C Monoclonal antibody specifically detects Tenascin C in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot.
Specifications

Specifications

Tenascin C
Monoclonal
8.3 mg/mL
Ascites with 15mM sodium azide
P24821
TNC
Purified, full length, native human protein.
RUO
3371
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot
BC-24
Unconjugated
TNC
150-225; AI528729; C130033P17Rik; cytotactin; deafness, autosomal dominant 56; DFNA56; Glioma-associated-extracellular matrix antigen; GMEM; GP; GP 150-225; hexabrachion; hexabrachion (tenascin); Hxb; JI; MGC144208; MGC144209; MGC167029; Myotendinous antigen; neuronectin; Ten; Tenascin; tenascin C; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; tnc; TN-C
Mouse
100 μL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions