Learn More
Tenascin R Monoclonal Antibody (1D12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA520139
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: VATHLSTPQG LQFKTITETT VEVQWEPFSF SFDGWEISFI PKNNEGGVIA QVPSDVTSFN QTGLKPGEEY IVNVVALKEQ ARSPPTSASV STVIDGPTQI LVRDVSDTV
Tenascin R Monoclonal antibody specifically detects Tenascin R in Human samples. It is validated for ELISA.Specifications
Tenascin R | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q92752 | |
Tnr | |
TNR (NP_003276, 411 a.a. ∽ 519 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
1D12 | |
Unconjugated | |
Tnr | |
J1NRM; J1-tenascin; janusin; Neural recognition molecule J1-160/180; restrictin; tenascin R; tenascin R (restrictin, janusin); tenascin-R; Tenascin-R (Restrictin janusin J1-160/180); Tenascin-R (Restrictin, janusin, J1-160/180); Tnr; TN-R | |
Mouse | |
Protein A | |
RUO | |
7143 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |