missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET2 Monoclonal Antibody (P411), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518299
Additional Details : Weight : 0.00910kg
Description
Recommended positive controls: DDDDK-human TET2-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TET2 Monoclonal antibody specifically detects TET2 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
TET2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q6N021 | |
TET2 | |
Recombinant fragment corresponding to a region within amino acids 429 and 637 of TET2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
P411 | |
Unconjugated | |
TET2 | |
Ayu17-449; E130014J05Rik; FLJ20032; KIAA1546; MDS; methylcytosine dioxygenase TET2; MGC125715; mKIAA1546; Nbla00191; OTTHUMP00000161869; probable methylcytosine dioxygenase TET2; Protein Ayu17 449; Protein Ayu17-449; RGD1311625; Tet 2; tet methylcytosine dioxygenase 2; tet oncogene 2; tet oncogene family member 2; TET2 | |
Mouse | |
Protein G | |
RUO | |
54790 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |