Learn More
TET2 Mouse anti-Human, Polyclonal, Invitrogen™
Mouse Polyclonal Antibody
Brand: Invitrogen PA535847
Additional Details : Weight : 0.00910kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.Specifications
TET2 | |
Polyclonal | |
Unconjugated | |
Q6N021 | |
TET2 | |
Recombit fragment corresponding to a region within amino acids 429 and 637 of TET2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay, Immunoprecipitation, Western Blot | |
0.88 mg/mL | |
TET2 | |
KIAA1546, MDS, tet methylcytosine dioxygenase 2 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
54790 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |