missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THC Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA175457
Additional Details : Weight : 0.01000kg
Description
PA1-75457 specifically detects Tetrahydrocannabinol. PA1-75457 has been successfully used in ELISA applications. The PA1-75457 immunogen is THC(9)-BSA.
THC Polyclonal antibody specifically detects THC in Chemical samples. It is validated for ELISA.Specifications
THC | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Sheep | |
purified | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
Unconjugated | |
C21H30O2; Delta9-Tetrahydrocannabinol; Delta9-THC; Deltanyne; Dronabinol; Marinol; Tetrahydrocannabinol | |
THC(9)-BSA | |
1 mg | |
Primary | |
Antibody | |
IgG |