missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518766
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
TFEB Polyclonal antibody specifically detects TFEB in Human samples. It is validated for Western Blot.Specifications
TFEB | |
Polyclonal | |
Unconjugated | |
TFEB | |
Alpha TCFEB; ALPHATFEB; BHLHE35; Class E basic helix-loop-helix protein 35; RP4-696P19.3; T-cell transcription factor EB; Tcfeb; Tfeb; Transcription factor EB | |
Goat | |
Antigen affinity chromatography | |
RUO | |
7942 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P19484 | |
TFEB | |
Synthetic peptide sequence (QKDLQKSRELENH) corresponding to the internal amino acids of TFEB | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |