missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544873
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: NANNFYTWEA CDDACWRIEK VPKVCRLQVS VDDQCEGSTE KYFFNLSSMT Sequence homology: Human: 100%
TFPI2 Polyclonal antibody specifically detects TFPI2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
TFPI2 | |
Polyclonal | |
Unconjugated | |
TFPI2 | |
AV000670; Placental protein 5; PP5; PP5/TFPI-2; PP5FLJ21164; REF1; REF-1; retinal pigment epithelium cell factor 1; TFPI2; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7980 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P48307 | |
TFPI2 | |
Synthetic peptide directed towards the middle region of human TFPI2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |