Learn More
TGF beta-1 Monoclonal Antibody (4F9C10,1B11B7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515289
Additional Details : Weight : 0.01000kg
Description
MA5-15289 targets TGF beta1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15289 immunogen is purified recombinant fragment of TGF beta1 expressed in E. Coli.
TGF beta-1 Monoclonal antibody specifically detects TGF beta-1 in Human samples. It is validated for ELISA, Western Blot.Specifications
TGF beta-1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P01137 | |
Tgfb1 | |
Purified recombinant fragment of TGF beta1 expressed in E. Coli. | |
RUO | |
7040 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
4F9C10,1B11B7, 1B11B7 | |
Unconjugated | |
Tgfb1 | |
CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF β; TGF β 1; TGF β1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFβ1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor β1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |