missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrxR1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518068
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with porcine based on sequence homology.
TrxR1 Polyclonal antibody specifically detects TrxR1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
TrxR1 | |
Polyclonal | |
Unconjugated | |
TXNRD1 | |
gene associated with retinoic and IFN-induced mortality 12 protein; gene associated with retinoic and interferon-induced mortality 12 protein; GRIM12; GRIM-12; KDRF; KM-102-derived reductase-like factor; MGC9145; NADPH-dependent thioredoxin reductase; oxidoreductase; selenoprotein oxidoreductase; testis tissue sperm-binding protein Li 46a; thioredoxin reductase 1; thioredoxin reductase 1, cytoplasmic; thioredoxin reductase GRIM-12; thioredoxin reductase TR1; TR; TR alpha; TR1; Trxr1; TXNR; Txnrd1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
7296 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q16881 | |
TXNRD1 | |
Synthetic peptide sequence (TKRSGASILQAGC) corresponding to the C-terminus amino acids of TXNRD1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |