Learn More
Thrombospondin 1 Monoclonal Antibody (A6.1), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513395
Additional Details : Weight : 0.01000kg
Description
MA5-13395 targets Thrombospondin in FACS, IF, IHC (P), IM, IP, and WB applications and shows reactivity with Bovine, Canine, Equine, Human, mouse, Ovine, Porcine, and Rat samples. The MA5-13395 immunogen is reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets.
Thrombospondin 1 Monoclonal antibody specifically detects Thrombospondin 1 in Bovine, Canine, Equine, Human, Mouse, Ovine, Porcine, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot.Specifications
Thrombospondin 1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P07996, P35441, Q28178 | |
THBS1 | |
Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets | |
500 μL | |
Primary | |
Bovine, Canine, Equine, Human, Mouse, Ovine, Porcine, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot | |
A6.1 | |
Biotin | |
THBS1 | |
glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 | |
Mouse | |
Protein G | |
RUO | |
100057478, 101106433, 21825, 281530, 445442, 487486, 492313, 7057 | |
4° C | |
Liquid |