Learn More
Thymidine Phosphorylase Monoclonal Antibody (P-GF.44C), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513542
Additional Details : Weight : 0.01000kg
Description
MA5-13542 targets Thymidine Phosphorylase in IHC (P) applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13542 immunogen is recombinant full length human thymidine phosphorylase (TP/PD-ECGF) protein.
Thymidine Phosphorylase Monoclonal antibody specifically detects Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Thymidine Phosphorylase | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P19971, Q5FVR2, Q99N42 | |
TYMP | |
Recombinant full length human thymidine phosphorylase (TP/PD-ECGF) protein | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
P-GF.44C | |
Unconjugated | |
TYMP | |
2900072D10Rik; ECGF; Ecgf1; endothelial cell growth factor 1 (platelet-derived); gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; thymidine phosphorylase; TP; TYMP | |
Mouse | |
Protein G | |
RUO | |
1890, 315219, 72962 | |
4° C | |
Liquid |