Learn More
Thyroid Stimulating Hormone Monoclonal Antibody (TSH-116), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA119067
Additional Details : Weight : 0.01000kg
Product Code. 11524502
  • 273.00€ / 100µg
Estimated Shipment: 12-04-2024
to see stock.
Description
Description
RIA: The antibody TSH-116 is suitable in combination with the antibody TSH-51 for immunometric assays in the screening of neonatal hypothyroidism.
Thyroid Stimulating Hormone Monoclonal antibody specifically detects Thyroid Stimulating Hormone in Human samples. It is validated for ELISA, Radioimmune Assays (RIA).
Specifications
Specifications
Thyroid Stimulating Hormone
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
P01215, P01222
CGA, TSHB
Human thyrotropin.
100 μg
Primary
Human
Antibody
IgG2a
ELISA, Radioimmune Assays (RIA)
TSH-116
Unconjugated
CGA
aGSU; alphaGSU; alpha-GSU; alphaSU; Anterior pituitary glycoprotein hormones common subunit alpha; Cga; Cga1; CG-alpha; Choriogonadotropin alpha chain; chorionic gonadotrophin subunit alpha; chorionic gonadotropin, alpha polypeptide; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin alpha chain; FSHA; FSH-alpha; Glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; glycoprotein hormones, alpha subunit; GPHa; GPHA1; GPHalpha; HCG; LHA; LSH-alpha; luteinizing hormone alpha chain; Lutropin alpha chain; pituitary hormone alpha subunit; thyroid stimulating hormone beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone alpha chain; Thyroid-stimulating hormone subunit beta; Thyrotropin; Thyrotropin alfa; Thyrotropin alpha chain; thyrotropin beta chain; Thyrotropin subunit beta; Tsha; TSH-alpha; Tshb; TSH-B; TSH-beta
Mouse
Protein A
RUO
1081, 7252
4° C, do not freeze