missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TNFRSF11A, Clone: 5A3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008792-M04.100ug
Additional Details : Weight : 0.02000kg
Specifications
TNFRSF11A | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003839.2 | |
TNFRSF11A | |
TNFRSF11A (NP_003830.1, 30 a.a. ∼ 212 a.a) partial recombinant protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
5A3 | |
Mouse monoclonal antibody raised against a partial recombinant TNFRSF11A. | |
TNFRSF11A | |
CD265/FEO/LOH18CR1/ODFR/OFE/OPTB7/OSTS/PDB2/RANK/TRANCER | |
Mouse | |
Affinity chromatography | |
RUO | |
8792 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
For Research Use Only