Learn More
anti-UBE2L6 Matched Antibody Pair, Abnova
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-UBE2L6 (100μg); Mouse purified polyclonal anti-UBE2L6 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is highly similar in primary structure to the enzyme encoded by UBE2L3 gene. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
ELISA
Specifications
Specifications
| Antigen | UBE2L6 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human UBE2L6. |
| Gene | UBE2L6 |
| Gene Alias | MGC40331/RIG-B/UBCH8 |
| Gene Symbols | UBE2L6 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.