missing translation for 'onlineSavingsMsg'
Learn More

Antide acetate, Thermo Scientific Chemicals

Catalog Number 15408149
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
112568-12-4
C82H108ClN17O14
1591.32
MFCD00133104,MFCD00133104
QRYFGTULTGLGHU-NBERXCRTSA-N
antide, iturelix, antide acetate, nal-lys-gnrh, unii-94055uoq3y, nal-lys-gnrhant, d-alaninamide, n-acetyl-3-2-naphthalenyl-d-alanyl-4-chloro-d-phenylalanyl-3-3-pyridinyl-d-alanyl-l-seryl-n6-3-pyridinylcarbonyl-l-lysyl-n6-3-pyridinylcarbonyl-d-lysyl-l-leucyl-n6-1-methylethyl-l-lysyl-l-prolyl, n-acetyl-3-naphhalen-2-yl-d-alanyl-p-chloro-d-phenylalanyl-3-3-pyridyl-d-alanyl-l-seryl-n sup 6-nicotinyl-l-lysyl-n sup 6-nicotinyl-d-lysyl-l-leucyl-n sup 6-isopropyl-l-lysyl-l-prolyl-d-alaninamide, d-alaninamide, n-acetyl-3-2-naphthalenyl-d-alanyl-4-chloro-d-phenylalanyl-3-3-pyridinyl-d-alanyl-l-seryl-n sup 6-3-pyridinylcarbonyl-l-lysyl-n sup 6-3-pyridinylcarbonyl-d-lysyl-l-leucyl-n sup 6-1-methylethyl-l-lysyl-l-prolyl, iturelix usan:inn
16130938
(2S)-N-[(1R)-1-{[(1S)-1-{[(2S)-1-[(2S)-2-{[(1R)-1-carbamoylethyl]carbamoyl}pyrrolidin-1-yl]-1-oxo-6-[(propan-2-yl)amino]hexan-2-yl]carbamoyl}-3-methylbutyl]carbamoyl}-5-[(pyridin-3-yl)formamido]pentyl]-2-[(2S)-2-[(2R)-2-[(2R)-3-(4-chlorophenyl)-2-[(2R)-2-acetamido-3-(naphthalen-2-yl)propanamido]propanamido]-3-(pyridin-3-yl)propanamido]-3-hydroxypropanamido]-6-[(pyridin-3-yl)formamido]hexanamide
CC(C)C[C@H](NC(=O)[C@@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CO)NC(=O)[C@@H](CC1=CC=CN=C1)NC(=O)[C@@H](CC1=CC=C(Cl)C=C1)NC(=O)[C@@H](CC1=CC=C2C=CC=CC2=C1)NC(C)=O)C(=O)N[C@@H](CCCCNC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@H](C)C(N)=O
This item is not returnable. View return policy
112568-12-4
C82H108ClN17O14
1591.32
MFCD00133104,MFCD00133104
QRYFGTULTGLGHU-NBERXCRTSA-N
antide, iturelix, antide acetate, nal-lys-gnrh, unii-94055uoq3y, nal-lys-gnrhant, d-alaninamide, n-acetyl-3-2-naphthalenyl-d-alanyl-4-chloro-d-phenylalanyl-3-3-pyridinyl-d-alanyl-l-seryl-n6-3-pyridinylcarbonyl-l-lysyl-n6-3-pyridinylcarbonyl-d-lysyl-l-leucyl-n6-1-methylethyl-l-lysyl-l-prolyl, n-acetyl-3-naphhalen-2-yl-d-alanyl-p-chloro-d-phenylalanyl-3-3-pyridyl-d-alanyl-l-seryl-n sup 6-nicotinyl-l-lysyl-n sup 6-nicotinyl-d-lysyl-l-leucyl-n sup 6-isopropyl-l-lysyl-l-prolyl-d-alaninamide, d-alaninamide, n-acetyl-3-2-naphthalenyl-d-alanyl-4-chloro-d-phenylalanyl-3-3-pyridinyl-d-alanyl-l-seryl-n sup 6-3-pyridinylcarbonyl-l-lysyl-n sup 6-3-pyridinylcarbonyl-d-lysyl-l-leucyl-n sup 6-1-methylethyl-l-lysyl-l-prolyl, iturelix usan:inn
16130938
(2S)-N-[(1R)-1-{[(1S)-1-{[(2S)-1-[(2S)-2-{[(1R)-1-carbamoylethyl]carbamoyl}pyrrolidin-1-yl]-1-oxo-6-[(propan-2-yl)amino]hexan-2-yl]carbamoyl}-3-methylbutyl]carbamoyl}-5-[(pyridin-3-yl)formamido]pentyl]-2-[(2S)-2-[(2R)-2-[(2R)-3-(4-chlorophenyl)-2-[(2R)-2-acetamido-3-(naphthalen-2-yl)propanamido]propanamido]-3-(pyridin-3-yl)propanamido]-3-hydroxypropanamido]-6-[(pyridin-3-yl)formamido]hexanamide
CC(C)C[C@H](NC(=O)[C@@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CO)NC(=O)[C@@H](CC1=CC=CN=C1)NC(=O)[C@@H](CC1=CC=C(Cl)C=C1)NC(=O)[C@@H](CC1=CC=C2C=CC=CC2=C1)NC(C)=O)C(=O)N[C@@H](CCCCNC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@H](C)C(N)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 mg
Formula Weight 1591.3
Physical Form Solid
Chemical Name or Material Antide acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'