missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ DNA Blunt End Kit
Click to view available options
Quantity:
100 reactions
Unit Size:
1 set
Description
Includes
Anza™ Blunting Enzyme Mix, Anza™ 10X Blunting Buffer,
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
- Part of the Anza™ Restriction Enzyme Cloning System
- Use directly following heat inactivation of digestion reaction
- Quick 15 minute protocol
Certifications
CoA
Specifications
Specifications
| Content And Storage | 100 μL Anza Blunting Enzyme Mix 200 μL Anza 10X Blunting Buffer Store at -5 to -30°C. |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Enzyme | T4 DNA polymerase, Klenow Fragment |
| Compatible Buffer | Anza™ 10X Blunting Buffer |
| Quantity | 100 reactions |
| Product Type | DNA Blunt End Kit |
| Product Line | Anza |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction