Learn More
Miele Professional™ Appliance lid
Product Code. 10098481 Shop All Miele Products
Click to view available options
:
Lid; Dimensions (D x W): 60 x 90mm
Product Code. 10098481
Brand: Miele Professional™ 5744710
Only null left
Add to basket
Add to basket
Appliance lid DE-CS6-78 for the top cover of machine incl. side unit, 60 cm deep, 90 cm wide
To cover the top of a machine
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Product Type Cover
Color Stainless Steel
For Use With (Equipment) Washer-disinfectors, laboratory