missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Apron

Product Code. 12689771 Shop All Ansell Products
Click to view available options
Dimensions (L x W):
33 x 43 in.
90 x 115 cm
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 12689771

Brand: Ansell Edmont™ CW45PT

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Type Apron
Material Vinyl
Thickness 0.5 mm
Includes Smooth edge laces on the sides.
Dimensions (L x W) 90 x 115 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.