Learn More
Julabo™ Aqua Stabil™ Water Bath Protective Media
Julabo™ Aqua Stabil™ Water Bath Protective Media stops growth of algae, bacteria and other microorganisms with its bactericidal effectiveness.
Brand:  Julabo™ 8940012
Additional Details : Weight : 1.48500kg
Product Code. 11930298
  • 417.00€ / Pack of 12
In Stock
to see stock.
Add to basket
Description
Description
  • Protects bath fluid
  • Provides long period of use for fluid
Specifications
Specifications
12 Bottles
Aqua Stabil Protective Media
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Aqua-stabil
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.