Learn More
Macherey-Nagel™ Aquadur™ Test Strips
Shop All Macherey-Nagel Products
Click to view available options
Graduations:
<3, >4, >7, >14, <21°
<3, >5, >10, >15, <20, >25°
Unit Size:
Pack of 100
Description
- Quick and easy determination of water hardness.
- The convenient dip and read procedure and the clear colour changes from green to red ensure reliable results within seconds.
- Water hardness test strips come in a wide variety of different gradations and packaging options for your individual needs.
Specifications
Specifications
| Certifications/Compliance | CE, 93/42 / EEC |
| Format | Strip |
| For Use With (Application) | Determination of water hardness |
| Graduations | <3, >4, >7, >14, <21° |
| Quantity | 100 Pack |
| Test Time | 20 sec. |
| Type | Aquadur Test Strips |