Learn More
Macherey Nagel Aquadur™ Water Hardness Test Strips
For semi-quantitative determination of water hardness (CaC03)
Brand:  Macherey Nagel 91224
Product Code. 12752721
  • ONLINE EXCLUSIVE PRICE
    306.00€
    296.00€ / Pack of 1000
    Save 10.00€
    3% Off
Only null left
In Stock
Add to basket
Description
Description
  • Test papers allow the semi-qualitative determination of water hardness
  • Ranged test results
Specifications
Specifications
CaCO3
1000 individually sealed strips
<54, >72, >26, >252, >375 ppm
Test strips, semi-quantitative, water hardness