missing translation for 'onlineSavingsMsg'
Learn More

Arbutin, 98%, Thermo Scientific Chemicals

Catalog Number p-7077569
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
497-76-7
C12H16O7
272.25
MFCD00016915
BJRNKVDFDLYUGJ-UHFFFAOYNA-N
arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin
440936
CHEBI:18305
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O
This item is not returnable. View return policy
497-76-7
C12H16O7
272.25
MFCD00016915
BJRNKVDFDLYUGJ-UHFFFAOYNA-N
arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin
440936
CHEBI:18305
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 195.0°C to 201.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: 50g/L. Other solubilities: soluble in alcohols
Formula Weight 272.25
Percent Purity 98%
Chemical Name or Material Arbutin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'