missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Arctic Gloves

Catalog Number p-7115897
Click to view available options
Size:
8
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Handle frozen products safely and comfortably. Honeywell Safety Products™ Arctic Gloves can be used in cold storage conditions and by forklift operators and drivers.

  • Grain goatskin.
  • Water repellant Scotchguard™ treatment.
  • Synthetic fur.
  • Elasticized on back with zig-zag stitching.
  • Good flexibility and outstanding dexterity.
  • Supple.
  • Good mechanical resistance.
TRUSTED_SUSTAINABILITY
Certifications/Compliance CE CATEGORY: II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 511-2004 Cold hazards, EC Category PPE: 2, Quality Assurance: ISO 9001/2000, EC Certificate Number: N°0075/797/162/07/12/0587, EC Attestation Number: N°0075/797/162/07/12/0587
Classification Dexterity: 5
Color Beige
Cut Resistance Rating 2
Material Leather
Product Type Gloves
Size 11
For Use With (Application) Agriculture, Building, Construction, Catering, Energy, Electricity, Food, Green Spaces, Iron, Steel, Logistics, Offshore, Ship Building, Telecom, Utility and Wood industries
Includes Elasticized back, Scotchguard™ treatment
Liner Material Synthetic fur
Lining Lined
Puncture Rating 1
Resistant To Cold
Tear Rating 2
Length (Metric) Cuff 30cm
Diameter (Metric) Cuff 10cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'