Learn More
Aalborg™ Argon Direct Glass Flow Tube
Product Code. p-8008814
Click to view available options
Airflow Rate:
1 L/min.
15 L/min.
2 L/min.
2.5 L/min.
25 L/min.
325 mL/min.
Width (Metric):
65 mm
150 mm
Unit Size:
Each
Product Code. 11892210
Brand: Aalborg™ FT02304GLVN
Only null left
Add to basket
Add to basket
Aalborg™ Argon Direct Glass Flow Tube completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Type Argon Direct Flow Tube
Width (Metric) 65 mm
Airflow Rate 2.5 L/min.