Learn More
Sartorius Arium™ Advance L Activated Carbon Filter
Arium™ Advance L Activated Carbon Filter effectively removes free chlorine and organic contamination.
Brand: Sartorius H2O-CACL-1
204.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- The Advance L activated carbon filter effectively removes free chlorine and organic contamination.
Specifications
Potable Tap Water | |
Reverse Osmosis (Type 3) | |
Filters | |
11 x 18 x 26 cm |