Description
The arium™ conversion kit extended tubing and its display and dispensing unit let you optimally integrate this equipment into your laboratory environment. This built-in version saves space on and above your lab bench as the control unit along with the display and dispenser permits variable wall-mounting options to suit your particular requirements.
Specifications
Specifications
| For Use With (Equipment) | Arium ProArium Comfort I |
| Type | Conversion Kit |
| For Use With (Application) | Water Purification |
| Material | PVDF |
| Width (Metric) | 3.4 m |