Learn More
WTW™ Armoring with Protective Hood for WTW™ ConOx™ and Tetracon™
The universal arming for the TetraCon™ 325 as well as for the galvanized oxygen sensor CellOx™ 325 provide optimal protection against damage in rough field applications, such as streaming water or bore holes.The special arming A pHLab/K is used to protect SenTix™ pH electrodes with glass shafts when measuring outside the lab.
Brand: WTW™ 903831
Additional Details : Weight : 0.80000kg
Description
- SensoLyt™ 900-P
- SensoLyt™ ORP 900-P
- TetraCon™ 925-P
- FDO™ 925-P
- VisoTurb™ 900-P
Specifications
Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325 | |
Armoring | |
Stainless Steel protective hood | |
Stainless Steel |