Learn More
Abeba™ Arrow 38400 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand: Abeba™ 38400/39
Additional Details : Weight : 0.60000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRA
- Smooth leather white
- Slip resistance PU/PU outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Microfiber heel patch
- Adjustable instep area
- velcro fastening
- Lining microfiber, respiration absorbing
Specifications
CE EN ISO 20347:2012 OB A, FO SRA | |
Microfiber | |
39 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
38400 |