Learn More
Abeba™ Arrow 38400 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand:  Abeba™ 38400/43
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 15671188
  • 81.83€ / 1 pair
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRA
  • Smooth leather white
  • Slip resistance PU/PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Microfiber heel patch
  • Adjustable instep area
  • velcro fastening
  • Lining microfiber, respiration absorbing
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRA
Microfiber
43
Polyurethane
White
Leather
Slip-resistant
38400