Learn More
Abeba™ Arrow 38600 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
85.73€
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRA
Color White
Liner Material Microfiber
Material Leather
Sole Features Slip-resistant
View More Specs
Products 12
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15631198
View Documents Promotion Details
Abeba™
38600/36
36
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15641198
View Documents Promotion Details
Abeba™
38600/37
37
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15651198
View Documents Promotion Details
Abeba™
38600/38
38
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15661198
View Documents Promotion Details
Abeba™
38600/39
39
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15671198
View Documents Promotion Details
Abeba™
38600/40
40
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15681198
View Documents Promotion Details
Abeba™
38600/41
41
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15691198
View Documents Promotion Details
Abeba™
38600/42
42
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15601208
View Documents Promotion Details
Abeba™
38600/43
43
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15611208
View Documents Promotion Details
Abeba™
38600/44
44
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15621208
View Documents Promotion Details
Abeba™
38600/45
45
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15631208
View Documents Promotion Details
Abeba™
38600/46
46
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15641208
View Documents Promotion Details
Abeba™
38600/47
47
85.73€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRA
  • Smooth leather, perforated
  • White
  • Slip resistance PU/PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Microfiber heel patch
  • Instep area padded with elastic band
  • velcro fastening
  • Lining microfiber, respiration absorbing
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRA
Microfiber
Slip-resistant
38600
White
Leather
Polyurethane