Learn More
Abeba™ Arrow 8212 Shoes
Product Code. 15663068
Click to view available options
Size:
48
49
50
51
Unit Size:
1 pair
Product Code. 15663068
Brand: Abeba™ 8212/51
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Antistatic with heel patch microfiber. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB A, E, FO SRA
  • Smooth leather
  • Black
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Leather lining
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRA
Color Black
Material Leather
Size 51
Sole Features Slip-resistant
Sole Material Polyurethane
Style 8212