Learn More
Abeba™ Arrow 8400 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand:  Abeba™ 8400/47
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 15663098
  • 79.22€ / 1 pair
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRA
  • Smooth leather white
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Microfiber heel patch
  • Adjustable instep area
  • Lining microfiber, respiration absorbing
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRA
Microfiber
47
Polyurethane
White
Leather
Slip-resistant
8400