missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) chloride, 99.5%, Thermo Scientific™

Product Code. 10114260 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10114260

Brand: Thermo Scientific Acros 190480100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 7784-34-1
Color Colorless to Yellow
Physical Form Liquid
Quantity 10 mL
Assay Percent Range 99.5%
Molecular Formula AsCl3
Linear Formula AsCl3
MDL Number MFCD00014165
Merck Index 15, 792
Synonym trichloroarsine, arsenic trichloride, arsenic iii chloride, arsenic chloride, arsenous trichloride, arsenic butter, arsenous chloride, arsenious chloride, butter of arsenic, chlorure arsenieux
InChI Key OEYOHULQRFXULB-UHFFFAOYSA-N
SMILES Cl[As](Cl)Cl
IUPAC Name trichloroarsane
Molecular Weight (g/mol) 181.272
PubChem CID 24570
ChEBI CHEBI:63952
Formula Weight 184.3
Percent Purity ≥99.4%
Packaging Glass bottle
Density 2.1500g/mL
Chemical Name or Material Arsenic(III) chloride, 99.5%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.