missing translation for 'onlineSavingsMsg'
Learn More

Arsenic (III) Standard: As+3 @ 100 μg/mL in 2% HCl, VHG™

Product Code. 17927819
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 17927819

Brand: VHG Labs VHGSPAS350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Arsenic (III) Standard: As+3 @ 100 μg/mL in 2% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arsenic+3
CAS 22541-54-4
Methods Speciation
Analyte or Component Names Arsenic+3
Shipping Condition Room Temperature
Molecular Formula As
Synonym As3+
InChI Formula InChI=1S/As/q+3
SMILES [As+3]
IUPAC Name arsenic(3+)
Solvent or Matrix 2% HCl
Molecular Weight (g/mol) 74.922
Quantity 50 mL
Formula Weight 74.922 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.