Description
- For use with AS 300 Sieve Shakers
Specifications
Specifications
| Product Type | Clamping Devices |
| Diameter (Metric) | 315 mm |
| For Use With (Application) | Dry Sieving |
| For Use With (Equipment) | Analytical sieve shaker AS 300 |
| Packaging Quantity | 1 Ea. |