Learn More
CPC (Colder) AseptiQuik™ G Genderless Quick-Disconnect Fittings, Hose Barb Adapter, Polycarbonate
Product Code. 30053910
Click to view available options
Diameter (English) Inner:
1/2 in.
1/4 in.
3/4 in.
3/8 in.
Includes:
With Valve
Without Valve
Unit Size:
Each
Product Code. 30053910
Brand: CPC (Colder) 5011154
Only null left
Add to basket
Add to basket
Genderless sterile connectors simplify process integration while maximizing flexibility
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Robust construction provides reliable and repeatable fluid transfer. 0.5 in. nominal flow path size
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/4 in.
Min. Operating Temperature (Metric) 1°C
Packaging Quantity 1 Ea.
Outlet Port Quick Disconnect x Hose Barb
Sterilization Method Gamma irradiation up to 50 kGy
Shape Straight
Includes Without Valve
Material (Seal) Platinum-cured Silicone
Color White with blue pull tab