missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATAGO™ Digital Hand-Held Pocket Refractometer, PAL-91S
Measure concentration and freezing point of Ethylene Glycol coolant in brine and other agents easily
Brand: ATAGO™ 4491
Additional Details : Weight : 0.10000kg
Description
- Pocket refractometer with digital display
- Battery powered, two AAA
- Measurement time of 3 seconds
Order Info
Optional storage case and NIST-Traceable™ Calibration Certificate available.
Specifications
0.0 to 90.0% (V/V) Concentration, 0°C to -50°C Freezing Point | |
0.3 mL or more | |
Conc.: ±0.4% (V/V), Freezing Point: ±1°C | |
Digital | |
Measuring concentration and freezing point of Ethylene Glycol coolant in brine and other agents | |
0.2 lb. | |
4.3 in. | |
2.1 in. | |
10°C to 40°C |
PAL-91S | |
Water only | |
Concentration: 0.2% (V/V); Freezing Point: 1°C | |
3 sec. | |
IP65 Water Resistant | |
100 g | |
109 mm | |
55 mm | |
2xAAA Batteries |