Learn More
ATAGO™ Master-M Refractometer
The MASTER-M series is a good alternative choice when the water resistant feature is not a priority and the Temperature Compensation will be carried out manually. The ATAGO™ Master-M Refractometer is not equipped with the Automatic Temperature Compensation feature.
Brand:  ATAGO™ 2313
Additional Details : Weight : 0.80000kg
Product Code. 10794367
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
The MASTER-M is made of metal; for salty and acidic samples, choose the plastic body model (MASTER-PM).
Specifications
Specifications
Refractometer
Master-M
Min. Scale ±0.2%
0.0 to 33.0 Brix
± 0.2%
Brix