Learn More
ATAGO™ Master-S28M Salinity Refractometer
The ATAGO™ Master-S28M Salinity Refractometer is a good alternative choice when the water resistant feature is not a priority and the Temperature Compensation will be carried out manually.
Brand:  ATAGO™ 2483
Additional Details : Weight : 0.16200kg
Product Code. 11466148
  • 252.00€ / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
  • Can be used for controlling the concentration of saltwater used in rinsing seafood or salinity in cooking
  • New material (which is durable for salty and acid sample) is adopted
Specifications
Specifications
Refractometer
Master-S28M
Sodium Chloride Percentage
0.0 to 28.0
Min. Scale ±0.2%