missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promed Bioscience Atelocollagen, Porcine Type I, Ultrapure (Enzymatic extraction, dialyzed)
Lyophilized, ultrapurified (via dialysis) Type I atelocollagen with superior functionality and physical/mechanical properties (strength, elasticity), suitable for bioprinting, cell culture, functional assays and in vivo research applications
198.90€ - 341.70€
Specifications
Formulation | Lyophilized Fibrous Powder |
---|---|
Purification Method | Double Dialysis |
Content And Storage | -20°C to 8°C |
Endotoxin Level | <1.0 EU/mL |
Purity or Quality Grade | >97% (by SDS/Western blotting) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17512845
|
Promed Bioscience
PRO-2003-10 |
10 mg |
198.90€
10mg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17522845
|
Promed Bioscience
PRO-2003-20 |
20 mg |
341.70€
20mg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Lyophilized, ultrapurified (via dialysis) Type I atelocollagen with superior functionality and physical/mechanical properties (strength, elasticity)
- The proprietary enzyme-based extraction removes telopeptides at the non-helical N and C termini, resulting in a triple helix configuration that exhibits reduced immunogenicity and enhanced biocompatibility
- Suitable as bioink for 3D bioprinting, for in vitro cell cultures, for cell functional assays and for in vitro and in vivo research applications (wound healing, tissue regeneration, drug delivery)
Specifications
Lyophilized Fibrous Powder | |
-20°C to 8°C | |
>97% (by SDS/Western blotting) |
Double Dialysis | |
<1.0 EU/mL |